CAS RN 1873364-08-9
CAS RN 1873364-08-9
IUPAC Name:
3,6-dibromo-9-(3,5-dimethylphenyl)-9H-carbazole
Synonyms:
3,6-dibromo-9-(3,5-dimethylphenyl)carbazole
Molecular Weight (g/mol):
429.16
Molecular Formula:
C20H15Br2N
InChi Key:
ASACIWHKQZICTN-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC(C)=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
1
–
1
of
1
results
1
3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole 97.0+%, TCI America™
CAS: 1873364-08-9 Molecular Formula: C20H15Br2N Molecular Weight (g/mol): 429.16 MDL Number: MFCD31618134 InChI Key: ASACIWHKQZICTN-UHFFFAOYSA-N PubChem CID: 129319319 IUPAC Name: 3,6-dibromo-9-(3,5-dimethylphenyl)-9H-carbazole SMILES: CC1=CC(=CC(C)=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2