accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1875-88-3

IUPAC Name: 2-(4-chlorophenyl)ethan-1-ol
Synonyms: 4-(2-hydroxyethyl)-chlorbenzol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: HZFRKZWBVUJYDA-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)