accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFOF

CAS RN 187543-87-9

IUPAC Name: 2,6-difluoro-3-methoxybenzaldehyde
Synonyms: 2,6-difluoro-3-methoxybenzaldehyde
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: WJCGNXHXQFFRKB-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C(C=O)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results