accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3CH3

CAS RN 18765-09-8

IUPAC Name: trioctylsilyl
Synonyms: trioctylsilyl
Molecular Weight (g/mol): 367.76
Molecular Formula: C24H51Si
InChi Key: QTKHQYWRGFZFHG-UHFFFAOYSA-N
SMILES: CCCCCCCC[Si](CCCCCCCC)CCCCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)