accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CHS

CAS RN 1878-18-8

IUPAC Name: 2-methylbutane-1-thiol
Synonyms: 2-methyl-1-butanethiol
Molecular Weight (g/mol): 104.21
Molecular Formula: C5H12S
InChi Key: WGQKBCSACFQGQY-UHFFFAOYNA-N
SMILES: CCC(C)CS
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)