accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1878-67-7

IUPAC Name: 2-(3-bromophenyl)acetic acid
Synonyms: M-bromophenylacetic acid
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: KYNNBXCGXUOREX-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)