accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1878-81-5

H3COOOHO

CAS RN 1878-81-5

IUPAC Name: 2-(4-acetylphenoxy)acetic acid
Synonyms: P-acetylphenoxyacetic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: KMXZEXUYXUMHEQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(OCC(O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results