accessibility menu, dialog, popup

UserName

Viewing profile as user:
Si

CAS RN 18784-61-7

IUPAC Name: diphenylbis(2-phenylethynyl)silane
Synonyms: diphenylbis(2-phenylethynyl)silane
Molecular Weight (g/mol): 384.55
Molecular Formula: C28H20Si
InChi Key: BBVOORFIXBLFFX-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C#C[Si](C#CC1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results