accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 18787-64-9

IUPAC Name: hexadecan-3-one
Synonyms: 3-hexadecanone
Molecular Weight (g/mol): 240.43
Molecular Formula: C16H32O
InChi Key: LTMXHUUHBSCKEK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)CC
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)