accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18791-02-1

IUPAC Name: 2,3-dibromopropanoyl chloride
Synonyms: 2,3-dibromopropanoyl chloride
Molecular Weight (g/mol): 250.31
Molecular Formula: C3H3Br2ClO
InChi Key: HWKWYDXHMQQDQJ-UHFFFAOYNA-N
SMILES: ClC(=O)C(Br)CBr
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)