accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHICl

CAS RN 188057-26-3

IUPAC Name: 6-chloro-2-iodopyridin-3-ol
Synonyms: 6-chloro-2-iodopyridin-3-ol
Molecular Weight (g/mol): 255.44
Molecular Formula: C5H3ClINO
InChi Key: FWIMPBYPMQSSCD-UHFFFAOYSA-N
SMILES: OC1=C(I)N=C(Cl)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)