accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18807-71-1

IUPAC Name: benzyl N-(2-aminoethyl)carbamate hydrochloride
Synonyms: benzyl N-(2-aminoethyl)carbamate hydrochloride
Molecular Weight (g/mol): 230.69
Molecular Formula: C10H15ClN2O2
InChi Key: QMLKQXIAPAAIEJ-UHFFFAOYSA-N
SMILES: Cl.NCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)