accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHNH2HCl

CAS RN 18807-73-3

IUPAC Name: benzyl N-(4-aminobutyl)carbamate hydrochloride
Synonyms: benzyl N-(4-aminobutyl)carbamate hydrochloride
Molecular Weight (g/mol): 258.75
Molecular Formula: C12H19ClN2O2
InChi Key: ZVNNCIIFBSRHFE-UHFFFAOYSA-N
SMILES: Cl.NCCCCNC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results