accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18812-51-6

IUPAC Name: diethyl propylphosphonate
Synonyms: diethyl propylphosphonate
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H17O3P
InChi Key: RUIKOPXSCCGLOM-UHFFFAOYSA-N
SMILES: CCCP(=O)(OCC)OCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results