accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHONH2

CAS RN 1882-72-0

IUPAC Name: methyl 2-amino-5-hydroxybenzoate
Synonyms: methyl 2-amino-5-hydroxybenzoate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DWBKSTKVIIRFHL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C=CC(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results