accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3O(E)

CAS RN 18829-55-5

IUPAC Name: (2E)-hept-2-enal
Synonyms: 2-heptenal, (2E)-
Molecular Weight (g/mol): 112.17
Molecular Formula: C7H12O
InChi Key: NDFKTBCGKNOHPJ-AATRIKPKSA-N
SMILES: CCCC\C=C\C=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)