CAS RN 18830-44-9
IUPAC Name:
methyl 3,3,3-trifluoropropanoate
Synonyms:
methyl 3,3,3-trifluoropropanoate
Molecular Weight (g/mol):
142.08
Molecular Formula:
C4H5F3O2
InChi Key:
PMGBATZKLCISOD-UHFFFAOYSA-N
SMILES:
COC(=O)CC(F)(F)F
1
–
1
of
1
results
1
Methyl 3,3,3-Trifluoropropionate 98.0+%, TCI America™
CAS: 18830-44-9 Molecular Formula: C4H5F3O2 Molecular Weight (g/mol): 142.077 MDL Number: MFCD02262206 InChI Key: PMGBATZKLCISOD-UHFFFAOYSA-N Synonym: 3,3,3-Trifluoropropionic Acid Methyl Ester PubChem CID: 2782392 IUPAC Name: methyl 3,3,3-trifluoropropanoate SMILES: COC(=O)CC(F)(F)F