accessibility menu, dialog, popup

UserName

Viewing profile as user:
S

CAS RN 1884-68-0

IUPAC Name: tetraphenylthiophene
Synonyms: tetraphenylthiophene
Molecular Weight (g/mol): 388.53
Molecular Formula: C28H20S
InChi Key: MQFBWJOMLIHUDY-UHFFFAOYSA-N
SMILES: S1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results