accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOO

CAS RN 1885-14-9

IUPAC Name: phenyl carbonochloridate
Synonyms: phenyl chloroformate
Molecular Weight (g/mol): 156.57
Molecular Formula: C7H5ClO2
InChi Key: AHWALFGBDFAJAI-UHFFFAOYSA-N
SMILES: ClC(=O)OC1=CC=CC=C1
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)