accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3COCH3N

CAS RN 1885-35-4

IUPAC Name: 3,4,5-trimethoxybenzonitrile
Synonyms: 3,4,5-trimethoxybenzonitrile
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: OSBQUSPVORCDCU-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C#N
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)