accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18856-63-8

IUPAC Name: 1-ethyl-4-isothiocyanatobenzene
Synonyms: 1-ethyl-4-isothiocyanatobenzene
Molecular Weight (g/mol): 163.24
Molecular Formula: C9H9NS
InChi Key: XXLWCGRHJFEMQH-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results