accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 188582-62-9

IUPAC Name: (4-bromo-2-fluorophenyl)methanol
Synonyms: (4-bromo-2-fluorophenyl)methanol
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: BWBJZMQPVBWEJU-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results