accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNH2NH2NOOHO

CAS RN 18860-78-1

IUPAC Name: N''-phenylguanidine; nitric acid
Synonyms: 2-phenylguanidine; nitric acid
Molecular Weight (g/mol): 198.18
Molecular Formula: C7H10N4O3
InChi Key: DIHKZQUSGDYVOT-UHFFFAOYSA-N
SMILES: O[N+]([O-])=O.NC(N)=NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results