CAS RN 188614-01-9
IUPAC Name:
methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Synonyms:
methyl 3-oxo-2,4-dihydro-1,4-benzothiazine-6-carboxylate
Molecular Weight (g/mol):
223.25
Molecular Formula:
C10H9NO3S
InChi Key:
UFSVRHNRNVVBJR-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC2=C(SCC(=O)N2)C=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, 97%
CAS: 188614-01-9 Molecular Formula: C10H9NO3S Molecular Weight (g/mol): 223.25 MDL Number: MFCD00449104 InChI Key: UFSVRHNRNVVBJR-UHFFFAOYSA-N PubChem CID: 699501 IUPAC Name: methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate SMILES: COC(=O)C1=CC2=C(SCC(=O)N2)C=C1