accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3(E)

CAS RN 18869-29-9

IUPAC Name: 1-methyl-4-[(1E)-2-(4-methylphenyl)ethenyl]benzene
Synonyms: 1-methyl-4-[(1E)-2-(4-methylphenyl)ethenyl]benzene
Molecular Weight (g/mol): 208.3
Molecular Formula: C16H16
InChi Key: KINZBJFIDFZQCB-VAWYXSNFSA-N
SMILES: CC1=CC=C(\C=C\C2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)