accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHBrNNOO

CAS RN 18874-52-7

IUPAC Name: 5-bromo-2-methyl-4-nitro-1H-imidazole
Synonyms: 4-bromo-2-methyl-5-nitro-3H-imidazole
Molecular Weight (g/mol): 206
Molecular Formula: C4H4BrN3O2
InChi Key: YOJYWZSEUWUYAQ-UHFFFAOYSA-N
SMILES: CC1=NC(=C(Br)N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results