accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18876-82-9

IUPAC Name: 2-methyl-1,3-thiazole-4-carboximidamide hydrochloride
Synonyms: 2-methyl-1,3-thiazole-4-carboximidamide hydrochloride
Molecular Weight (g/mol): 177.65
Molecular Formula: C5H8ClN3S
InChi Key: YYLYRAUCGUIGIE-UHFFFAOYSA-N
SMILES: Cl.CC1=NC(=CS1)C(N)=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results