accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3Br

CAS RN 18880-00-7

IUPAC Name: 1-(bromomethyl)-4-tert-butylbenzene
Synonyms: 1-(bromomethyl)-4-tert-butylbenzene
Molecular Weight (g/mol): 227.15
Molecular Formula: C11H15Br
InChi Key: QZNQSIHCDAGZIA-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CBr)C=C1
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (5)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)