accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18880-05-2

IUPAC Name: [(3-methoxyphenyl)methyl]triphenylphosphanium chloride
Synonyms: [(3-methoxyphenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 418.9
Molecular Formula: C26H24ClOP
InChi Key: DPYDLIVUYPUXBV-UHFFFAOYSA-M
SMILES: [Cl-].COC1=CC=CC(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results