accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFF

CAS RN 188815-30-7

IUPAC Name: 3-fluoro-5-(trifluoromethyl)benzaldehyde
Synonyms: 3-fluoro-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: UZZYXZWSOWQPIS-UHFFFAOYSA-N
SMILES: FC1=CC(C=O)=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results