accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CBrNH3CCH3CH3CH3FPFFFFF

CAS RN 188944-77-6

IUPAC Name: bis(2,4,6-trimethylpyridin-1-ium-1-yl)bromanuide; hexafluoro-λ⁵-phosphanuide
Synonyms: bis(2,4,6-trimethylpyridin-1-ium-1-yl)bromanuide hexafluorophosphate
Molecular Weight (g/mol): 467.23
Molecular Formula: C16H22BrF6N2P
InChi Key: YTYGAGSIUXQVJI-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CC1=CC(C)=[N+]([Br-][N+]2=C(C)C=C(C)C=C2C)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results