accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOPOOH3COH3C

CAS RN 188945-41-7

IUPAC Name: ethyl 2-[bis(2-methylphenoxy)phosphoryl]acetate
Synonyms: ethyl 2-[bis(2-methylphenoxy)phosphoryl]acetate
Molecular Weight (g/mol): 348.34
Molecular Formula: C18H21O5P
InChi Key: BOEGWWFFVZDOIH-UHFFFAOYSA-N
SMILES: CCOC(=O)CP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results