accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 189-55-9

IUPAC Name: hexacyclo[10.10.2.0²,⁷.0⁹,²³.0¹⁴,¹⁹.0²⁰,²⁴]tetracosa-1(22),2,4,6,8,10,12,14,16,18,20,23-dodecaene
Synonyms: dibenzo(A,I)pyrene
Molecular Weight (g/mol): 302.38
Molecular Formula: C24H14
InChi Key: TUGYIJVAYAHHHM-UHFFFAOYSA-N
SMILES: C1=CC=C2C(=C1)C=C1C=CC3=CC4=CC=CC=C4C4=CC=C2C1=C34
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)