accessibility menu, dialog, popup

UserName

Viewing profile as user:
II

CAS RN 189105-78-0

IUPAC Name: 1,8-diiodoanthracene
Synonyms: 1,8-diiodoanthracene
Molecular Weight (g/mol): 430.03
Molecular Formula: C14H8I2
InChi Key: BBKOQZVGDMYPGV-UHFFFAOYSA-N
SMILES: IC1=CC=CC2=CC3=CC=CC(I)=C3C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results