accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18911-76-7

IUPAC Name: yttrium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Synonyms: yttrium(3+) tris(1,1,1,5,5,5-hexafluoro-2,4-dioxopentan-3-ide)
Molecular Weight (g/mol): 710.06
Molecular Formula: C15H3F18O6Y
InChi Key: AJVDSGQQIVVYRX-UHFFFAOYSA-N
SMILES: [Y+3].FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F.FC(F)(F)C(=O)[CH-]C(=O)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results