accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18912-37-3

IUPAC Name: [(4-methoxyphenyl)methoxy]carbohydrazide
Synonyms: (4-methoxyphenyl)methoxycarbohydrazide
Molecular Weight (g/mol): 196.21
Molecular Formula: C9H12N2O3
InChi Key: JKBMMHKEAGEILO-UHFFFAOYSA-N
SMILES: COC1=CC=C(COC(=O)NN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results