CAS RN 18917-91-4
CAS RN 18917-91-4
IUPAC Name:
bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate
Synonyms:
aluminum lactate aluminium trilactate tris(lactato)aluminium
Molecular Weight (g/mol):
294.19
Molecular Formula:
C9H15AlO9
InChi Key:
VXYADVIJALMOEQ-UHFFFAOYNA-K
SMILES:
CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O
1
–
1
of
1
results
1
Aluminum lactate, 90-95%, contains ^=5% water
CAS: 18917-91-4 Molecular Formula: C9H15AlO9 Molecular Weight (g/mol): 294.19 MDL Number: MFCD20529175 InChI Key: VXYADVIJALMOEQ-UHFFFAOYNA-K IUPAC Name: bis[(2-hydroxypropanoyl)oxy]alumanyl 2-hydroxypropanoate SMILES: CC(O)C(=O)O[Al](OC(=O)C(C)O)OC(=O)C(C)O