accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18917-96-9

CH3OOHOCH3OOHOH2OH2OH2OMg

CAS RN 18917-96-9

IUPAC Name: magnesium(2+) bis(2-hydroxypropanoate) trihydrate
Synonyms: magnesium(2+) trihydrate dilactate
Molecular Weight (g/mol): 256.49
Molecular Formula: C6H16MgO9
InChi Key: SNLQXUYQWUDJLB-UHFFFAOYNA-L
SMILES: O.O.O.[Mg++].CC(O)C([O-])=O.CC(O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results