accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClNH2ONHOFNH

CAS RN 1893-33-0

IUPAC Name: 4'-carbamoyl-1'-[4-(4-fluorophenyl)-4-oxobutyl]-[1,4'-bipiperidine]-1,1'-diium dichloride
Synonyms: pipamperone
Molecular Weight (g/mol): 448.4
Molecular Formula: C21H32Cl2FN3O2
InChi Key: BMXXSXQVMCXGJM-UHFFFAOYSA-N
SMILES: [Cl-].[Cl-].NC(=O)C1(CC[NH+](CCCC(=O)C2=CC=C(F)C=C2)CC1)[NH+]1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results