accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOHNH3CH3CCH3

CAS RN 18934-81-1

IUPAC Name: 12-{[(tert-butoxy)carbonyl]amino}dodecanoic acid
Synonyms: 12-[(tert-butoxycarbonyl)amino]dodecanoic acid
Molecular Weight (g/mol): 315.45
Molecular Formula: C17H33NO4
InChi Key: NNPOZNVKUHYVEJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)