accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 189367-54-2

IUPAC Name: 2,7-dibromo-9,9-dihexyl-9H-fluorene
Synonyms: 2,7-dibromo-9,9-dihexylfluorene
Molecular Weight (g/mol): 492.34
Molecular Formula: C25H32Br2
InChi Key: OXFFIMLCSVJMHA-UHFFFAOYSA-N
SMILES: CCCCCCC1(CCCCCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)