accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH(R)OHOOOH3CCH3H3C

CAS RN 18942-49-9

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]-3-phenylpropanoic acid|BOC-D-PHE-OH
Molecular Weight (g/mol): 265.31
Molecular Formula: C14H19NO4
InChi Key: ZYJPUMXJBDHSIF-LLVKDONJSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (6)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)