accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18958-57-1

IUPAC Name: nickel(2+) bis((1,2-dicyano-2-sulfanidyleth-1-en-1-yl)sulfanide) bis(tetrabutylazanium)
Synonyms: nickel(2+) bis((1,2-dicyano-2-sulfanidyleth-1-en-1-yl)sulfanide) bis(tetrabutylammonium ion)
Molecular Weight (g/mol): 823.99
Molecular Formula: C40H72N6NiS4
InChi Key: KFDRXQXGGQXFNP-UHFFFAOYSA-J
SMILES: [Ni++].[S-]C(C#N)=C([S-])C#N.[S-]C(C#N)=C([S-])C#N.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results