accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1897-41-2

IUPAC Name: 1-[4-(aminomethyl)-2,3,5,6-tetrachlorophenyl]methanamine
Synonyms: 1-[4-(aminomethyl)-2,3,5,6-tetrachlorophenyl]methanamine
Molecular Weight (g/mol): 273.97
Molecular Formula: C8H8Cl4N2
InChi Key: SEDAWQAAXOJIPH-UHFFFAOYSA-N
SMILES: NCC1=C(Cl)C(Cl)=C(CN)C(Cl)=C1Cl
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results