accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNClN

CAS RN 1897-43-4

IUPAC Name: 2,5-dichlorobenzene-1,4-dicarbonitrile
Synonyms: 2,5-dichlorobenzene-1,4-dicarbonitrile
Molecular Weight (g/mol): 197.02
Molecular Formula: C8H2Cl2N2
InChi Key: UCRQGBRIGDKUAM-UHFFFAOYSA-N
SMILES: ClC1=CC(C#N)=C(Cl)C=C1C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results