accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18987-71-8

IUPAC Name: (chloraniumylpalladio)chloranium bis(2-[(dimethylamino)methyl]-4,6-dimethoxybenzen-1-ide) palladium
Synonyms: (chloraniumylpalladio)chloranium bis(2-[(dimethylamino)methyl]-4,6-dimethoxybenzen-1-ide) palladium
Molecular Weight (g/mol): 674.26
Molecular Formula: C22H34Cl2N2O4Pd2
InChi Key: KMCZAHZAAFWMFF-UHFFFAOYSA-N
SMILES: [Pd].[ClH+][Pd][ClH+].COC1=CC(CN(C)C)=[C-]C(OC)=C1.COC1=CC(CN(C)C)=[C-]C(OC)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results