accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOO

CAS RN 1899-24-7

IUPAC Name: 5-bromofuran-2-carbaldehyde
Synonyms: 5-bromofuran-2-carbaldehyde
Molecular Weight (g/mol): 174.98
Molecular Formula: C5H3BrO2
InChi Key: WJTFHWXMITZNHS-UHFFFAOYSA-N
SMILES: BrC1=CC=C(O1)C=O
Boiling Point 
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)