accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1899-48-5

IUPAC Name: quinazoline-2,4-diamine
Synonyms: 2,4-diaminoquinazoline
Molecular Weight (g/mol): 160.18
Molecular Formula: C8H8N4
InChi Key: XELRMPRLCPFTBH-UHFFFAOYSA-N
SMILES: NC1=NC(N)=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results