CAS RN 18996-35-5
Citric acid, monosodium salt, 99%, for analysis, anhydrous
CAS: 18996-35-5 Molecular Formula: C6H7NaO7 Molecular Weight (g/mol): 214.11 InChI Key: SLWOXBQHKZPUNY-UHFFFAOYSA-N Synonym: bicitra,pneucid,1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt,sodium dihydrogen citrate anhydrous,citric acid, sodium salt,citric acid sodium,sodium 2-hydroxy-1,2,3-propanetricarboxylate,2-hydroxypropane-1,2,3-tricarboxylic acid; sodium,1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:? PubChem CID: 131675399 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium SMILES: [HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]
Sodium dihydrogen citrate, 99%
CAS: 18996-35-5 Molecular Formula: C6H10NaO7 Molecular Weight (g/mol): 217.129 MDL Number: MFCD00013067 InChI Key: SLWOXBQHKZPUNY-UHFFFAOYSA-N Synonym: bicitra,pneucid,1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt,sodium dihydrogen citrate anhydrous,citric acid, sodium salt,citric acid sodium,sodium 2-hydroxy-1,2,3-propanetricarboxylate,2-hydroxypropane-1,2,3-tricarboxylic acid; sodium,1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:? PubChem CID: 131675399 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium SMILES: [HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]
Citric acid, monosodium salt, 99%, pure, anhydrous
CAS: 18996-35-5 Molecular Formula: C6H7NaO7 Molecular Weight (g/mol): 214.11 MDL Number: MFCD00013067 InChI Key: SLWOXBQHKZPUNY-UHFFFAOYSA-N Synonym: bicitra,pneucid,1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt,sodium dihydrogen citrate anhydrous,citric acid, sodium salt,citric acid sodium,sodium 2-hydroxy-1,2,3-propanetricarboxylate,2-hydroxypropane-1,2,3-tricarboxylic acid; sodium,1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:? PubChem CID: 131675399 IUPAC Name: 2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium SMILES: [HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]
Buffer concentrate pH 4.00, For 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution, Solstice
for 500 ml buffer solution, citric acid / sodium hydroxide / sodium chloride solution
Sigma Aldrich Sodium citrate monobasic
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Sodium dihydrogencitrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.