accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 19008-71-0

IUPAC Name: 8-aminooctan-1-ol
Synonyms: 8-aminooctan-1-ol
Molecular Weight (g/mol): 145.25
Molecular Formula: C8H19NO
InChi Key: WDCOJSGXSPGNFK-UHFFFAOYSA-N
SMILES: NCCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results